Learn More
Medchemexpress LLC (+)-Dibenzoyl-D-tartaric acid | 17026-42-5 | 99.8% | 358.30 | 500 G
SDP
Supplier:  Medchemexpress LLC HY20551500G
Encompass_Preferred
(+)-Dibenzoyl-D-tartaric acid is a biochemical reagent used as a biological material or organic compound for life science-related research. It is used as a reagent in the preparation of chiral salts and for chiral resolution of amino compounds. It is suitable for research use only.
  • Used as a biological material or organic compound for life science-related research
  • Reagent for preparing chiral salts
  • Used for chiral resolution of amino compounds
Catalog No. 50-004-00989