Learn More
Selleck Chemical LLC Dibenzoyl Thiamine S5474-25mg
SDP
Supplier:  Selleck Chemical LLC S547425MG
Encompass_Preferred
Dibenzoyl Thiamine (Bentiamine O S-Dibenzoylthiamine) a lipophilic derivative of vitamin B (thiamine) is a kind of food additive that can be rapidly absorbed into the body and converted to thiamine
Catalog No. 50-310-9997