Learn More
Medchemexpress LLC Dibenzoyl Thiamine (Bentiamine) | 299-88-7 | 99.8% | 490.57 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHYB2212
Encompass_Preferred
Dibenzoyl Thiamine (Bentiamine) is a derivative of thiamine that is rapidly absorbed into the body and subsequently converted to thiamine. It is a thiol-type thiamine composed of thiamine and benzoic acid. A key feature is that it is not decomposed by aneurinase (a thiamine-decomposing enzyme), making it more suitable for food processing compared to thiamine hydrochloride.
  • Rapid absorption into the body.
  • Converted to thiamine after absorption.
  • Not broken down by thiamine-decomposing enzymes, enhancing its suitability for food processing.
  • Chronic toxicity studies in male Wistar rats showed no specific toxic effects.
  • Teratogenicity studies in rats indicated no embryotoxic or teratogenic effects.
  • Mutagenicity tests revealed no mutagenic effects.
  • Metabolized to Vitamin B1 and benzoic acid, with benzoic acid excreted as hippuric acid.
Catalog No. 50-003-64984