Learn More
Medchemexpress LLC HY-W009731 500mg Medchemexpress, Dibenzoylmethane CAS:120-46-7 Purity:>98%
Supplier: Medchemexpress LLC HYW009731500MG
Medchemexpress, HY-W009731 500mg Dibenzoylmethane CAS:120-46-7 Dibenzoylmethane, a minor ingredient in licorice, activates Nrf2 and prevents various cancers and oxidative damage. Dibenzoylmethane, an analog of curcumin, results in dissociation from Keap1 and nuclear translocation of Nrf2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.