Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibenzyl aspartate 4-methylbenzenesulfonate | 4079-62-3 | MFCD00187195 | ≥95.0% | 485.55 | C25H27NO7S | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW092107

Encompass_Preferred

Dibenzyl aspartate 4-methylbenzenesulfonate is an aspartic acid derivative supplied for research use and not for clinical or human use.

  • Aspartic acid derivative
  • For research use only
  • Purity ≥95.0%
  • Molecular weight 485.55, molecular formula C25H27NO7S
  • Available in 25 g and 100 g sizes

Catalog No. 50-002-08749


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.