Learn More
AdipoGen Dibenzylfluorescein (100 mg)
SDP
Supplier:  AdipoGen CDXD0282M100
Encompass_Preferred
Chemical. CAS: 97744-44-0. Formula: C34H24O5. MW: 512.55. Dibenzylfluorescein is a fluorogenic probe that acts as a substrate for specific cytochrome P450 (CYP) isoforms, including CYP3A4, CYP2C8, CYP2C9, CYP2C19 and aromatase (CYP19). It is dealkylated by these CYP isoforms to produce fluorescein benzyl ether, which is further hydrolyzed to fluorescein by the addition of base (typically 2 M NaOH). Dibenzylfluorescein is typically used near its apparent Km value of 0.87-1.9 µM. The fluorescence of fluorescein is evaluated using excitation/emission wavelengths of 485/538nm. Dibenzylfluorescein is used to detect changes in CYP catalytic activity caused by drugs or disease.
Catalog No. 50-205-4380