Learn More
Apexbio Technology LLC Dibromoacetic acid 631-64-1 100mg
SDP
Supplier:  Apexbio Technology LLC A4740100
Encompass_Preferred
Dibromoacetic acid (CAS 631-64-1) is a halogenated acetic acid encountered as a disinfection byproduct in drinking water In biomedical studies it modulates immune responses and induces apoptosis through inhibition of specific cellular signaling pathways Research demonstrates its direct impact on the activity of certain enzymes relevant to immunological processes and programmed cell death Owing to these mechanistic roles Dibromoacetic acid is frequently utilized in studies investigating immunotoxicity and the cellular regulation associated with immune-mediated conditions
Catalog No. 50-000-95098