Learn More
Apexbio Technology LLC Dibucaine 85-79-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B33735.1
Encompass_Preferred
Dibucaine (CAS 85-79-0) is an aminoamide local anesthetic known to inhibit rapid axonal transport of proteins In in vitro assays Dibucaine at 0 5 mM exhibited a level of transport blockade comparable to 1 mM tetracaine while 1 mM Dibucaine reduced radiolabeled protein accumulation at ligation sites by 92% and 0 5 mM by 72% These properties underline its utility in studies investigating the mechanisms underlying axonal transport and the effects of local anesthetics on neuronal function
Catalog No. 50-000-96424