Learn More
Medchemexpress LLC Dibucaine | 85-79-0 | MFCD00047595 | 99.8% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB055210G
Encompass_Preferred
Dibucaine (Cinchocaine) is a sodium channel inhibitor and a potent SChE inhibitor, intended for research use only.
  • Reduces the degradation of BSA-gold complex in reservosomes.
  • Inhibits SChE within 2 minutes in a reversible manner.
  • Exhibits competitive inhibition with respect to BuTch (K(i) of 1.3 μM).
  • Shows linear-mixed type inhibition with respect to AcTch (K(i) of 0.66 μM and K(I) of 2.5 μM).
  • Contains a butoxy side chain.
Catalog No. 50-004-07888