Learn More
Apexbio Technology LLC Dibucaine (Cinchocaine) HCl 61-12-1 5g
SDP
Supplier:  Apexbio Technology LLC A84075000
Encompass_Preferred
Dibucaine (Cinchocaine) HCl (CAS 61-12-1) is a synthetic amide-type local anesthetic widely used in biomedical research It acts by reversibly binding to and inhibiting voltage-gated sodium channels in neuronal membranes thereby suppressing the initiation and propagation of nerve impulses Through this mechanism dibucaine impedes sensory signal transmission making it a valuable tool for investigating ion channel function neuronal excitability and pathways underlying pain perception Its application extends to in vitro studies of nerve conduction and as a reference compound in anesthetic pharmacology
Catalog No. 50-000-96036