Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibutyl phenyl phosphate | 2528-36-1 | MFCD00438060 | >95.0% | 286.30 | C14H23O4P | 25 MG
SDP

Supplier:  Medchemexpress LLC HYW18533625MG

Encompass_Preferred

Dibutyl phenyl phosphate is an organophosphate reagent supplied for research use, commonly used as a flame retardant and studied as a butyrylcholinesterase (BChE) inhibitor (IC50 83.6 μM; Ki 94.5 μM). It is provided in small laboratory quantities and identified by CAS 2528-36-1.

  • Flame retardant organophosphate.
  • BChE inhibitor (IC50 83.6 μM; Ki 94.5 μM).
  • Molecular formula C14H23O4P; molecular weight 286.30.
  • Available in small research quantities such as 25 MG.
  • Purity reported >95.0% (HPLC) by standards provider.
  • Intended for research use only; refer to SDS and CoA for handling.

Catalog No. 50-002-81584


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.