Learn More
Medchemexpress LLC Dibutyl phthalate | 84-74-2 | MFCD00009441 | 99.9% | 278.34 | C16H22O4 | 100mg
SDP
Supplier:  Medchemexpress LLC HYY0304100MG
Encompass_Preferred
Dibutyl phthalate is a commonly used plasticizer commonly found in some food packaging materials personal care products and the coating of oral medications[1] May cause toxicity and adverse neurobehavioral effects[2][3]
Catalog No. 50-002-68326