Learn More
Enzo Life Sciences Dibutyryl-cAMP (30mg). CAS: 16980-89-5
SDP
Supplier:  Enzo Life Sciences BMLCN1250030
Encompass_Preferred
A cell-permeable cAMP analog which preferentially activates cAMP-dependent protein kinase. The products releaes butyrate due to intracellular and extracellular esterase action. Butyrate was shown to have distinct biological effects. Alternative name: Dibutyl cyclic AMP sodium salt. Purity: ≥98% (HPLC). Solubility: Soluble in water (100mg/ml), DMSO (200mM) or 96% ethanol (25mM). Long Term Storage: -20°C.
Catalog No. 50-201-0442