Learn More
Selleck Chemical LLC Dibutyryl-cAMP (Bucladesine) 500mg 16980-89-5 dbcAMP
SDP
Supplier:  Selleck Chemical LLC S7858500MG
Encompass_Preferred
Dibutyryl-cAMP (Bucladesine, dbcAMP) is a cell-permeable PKA activator by mimicing the action of endogenous cAMP. Dibutyryl-cAMP (Bucladesine) is also a phosphodiesterase (PDE) inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6399