Learn More
STEMCELL Technologies Dibutyryl-cAMP (Sodium Salt), Size: 250 mg
SDP
Supplier:  STEMCELL Technologies 73886
Encompass
Dibutyryl-cAMP is a cell-permeable cyclic AMP (cAMP) analog that activates cAMP-dependent protein kinases (Schwede et al.). This product is supplied as the sodium salt of the molecule.DIFFERENTIATION· Suppresses experimental autoimmune encephalomyelitis development by reducing demyelination and mobilizing neural stem cells in the subventricular zone towards the demyelinated plaques (Khezri et al.).· Induces intrinsic axon growth in peripheral and central nervous systems, and morphological differentiation of astrocytes (Knott et al.; Imamura & Ozawa).· Stimulates neurite outgrowth in PC12 cells (Maruoka et al.).
Catalog No. 50-197-6142
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov