Learn More
Abcam Dibutyryl-cGMP, cGMP analog, 10MG
Supplier:  Abcam AB14520310MG
Encompass_Preferred
MW 507.4 Da, Purity >96%. Cell-permeable cGMP analog. Activates Protein Kinase G (PKG). Raises intracellular Ca²+ concentrations similar to 8-Br-cGMP (ab141449). Shows antinociceptive effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5399