Learn More
Cayman Chemical DIbutyrylCyclIc GMPsodIum 10mg
SDP
Supplier:  Cayman Chemical 1599110 MG
Encompass_Preferred
A cell-permeable, cGMP analog that activates cGMP-dependent protein kinase
Catalog No. 50-297-1815