Learn More
Enzo Life Sciences DICA (25mg). CAS: 613665-26-2
SDP
Supplier:  Enzo Life Sciences ALX420036M025
Encompass_Preferred
Inhibitor of caspase-3 and caspase-7 binding to an allosteric site and not to the active site. Alternative name: 2-(2,4-Dichlorophenoxy)-N-(2-mercaptoethyl)acetamide. Purity: ≥98% (TLC). Appearance: White to off-white solid. Solubility: Soluble in methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-9114