Learn More
Medchemexpress LLC Dicamba | 1918-00-9 | 100.0% | 25 G
SDP
Supplier:  Medchemexpress LLC HY12126725G
Encompass_Preferred
Dicamba is a chlorinated benzoic acid herbicide supplied as a high-purity small-molecule research reagent used in agricultural and environmental studies. It is provided with lot-specific quality and safety documentation for laboratory use.
  • Chlorinated benzoic acid (C8H6Cl2O3) with CAS 1918-00-9.
  • High purity suitable for analytical and research use.
  • Available in multiple pack sizes including 25 G for small-scale studies.
  • Supplied with COA and SDS for lot-specific quality and safety information.
  • Useful as a reference standard in herbicide research and environmental testing.
Catalog No. 50-002-33783