Learn More
Medchemexpress LLC (-)-Dicentrine | 28832-07-7 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN21051MG
Encompass_Preferred
(-)-Dicentrine is an aporphine alkaloid isolated from the stem bark of Talauma arcabucoana. It demonstrates moderate growth inhibition against Staphylococcus aureus and Candida albicans. Additionally, it exhibits cytotoxicity against various human and mouse cell lines after 3 days by SRB assay.
  • Isolated from the stem bark of Talauma arcabucoana.
  • Demonstrates moderate growth inhibition against Staphylococcus aureus and Candida albicans.
  • Exhibits cytotoxicity against various human and mouse cell lines.
  • For research use only.
Catalog No. 50-004-21978