Learn More
Strem, An Ascensus Company CAS# 50982-12-2. 1g. Dichloro(1,5-cyclooctadiene)ruthenium(II) polymer, min. 97%. MFCD00171304
SDP
Supplier:  Strem, An Ascensus Company 4404251G
Encompass
CAS# 50982-12-2. 1g. Dichloro(1,5-cyclooctadiene)ruthenium(II) polymer, min. 97%. MFCD00171304. Molecular Weight: 280.16. Molecular Formula: [Ru(1,5-C8H12)Cl2]x. Color/form: brown pwdr. Strem# 44-0425. http://www.strem.com/catalog/v/44-0425/
Catalog No. 50-901-11919