Learn More
Medchemexpress LLC Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer | 12354-84-6 | MFCD00075435 | ≥99.0% | 796.70 g·mol⁻¹ | C20H30Cl4Ir2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW018845100MG
Encompass_Preferred
[Ir(Cp-)Cl2]2 (dichloro(pentamethylcyclopentadienyl)iridium(III) dimer) is an organometallic iridium complex supplied for research use. It serves as a precursor to catalysts for asymmetric transfer hydrogenation of ketones and is employed in catalytic amine synthesis. The compound is a solid, typically orange to reddish brown, with formula C20H30Cl4Ir2 and an approximate molecular weight of 796.70 g·mol⁻¹.
  • High purity ≥99.0% suitable for research and synthetic applications.
  • Precursor for asymmetric transfer hydrogenation catalysts.
  • Useful catalyst for greener amine synthesis and related transformations.
  • Orange to reddish brown solid offering easy handling and storage.
  • Chemical identity confirmed by CAS number and molecular formula for traceability.
Catalog No. 50-003-01090