Learn More
Medchemexpress LLC Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer | 12354-85-7 | MFCD00061552 | 98.2% | 618.08 g·mol^-1 | C20H30Cl4Rh2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW019734100MG
Encompass_Preferred
[Rh(Cp-)Cl2]2 (pentamethylcyclopentadienylrhodium(III) chloride dimer) is an organometallic reagent provided as a solid for research use. It functions as a catalyst or catalyst precursor in a wide range of synthetic transformations, including hydrogenation, reductive alkylation, oxidative olefination, amidation, and C-C bond cleavage; it is typically handled under inert atmosphere and supplied with analytical documentation.
  • High purity solid suitable for catalytic applications.
  • Effective catalyst for hydrogenation and chiral hydrogenation.
  • Promotes oxidative olefination and oxidative annulation reactions.
  • Enables C-C bond cleavage of secondary alcohols in synthesis.
  • Supplied with COA and analytical data for quality verification.
Catalog No. 50-002-99582