Learn More
Medchemexpress LLC Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer | 12354-85-7 | MFCD00061552 | 98.2% | 618.08 g·mol⁻¹ | C20H30Cl4Rh2 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW019734
[Rh(Cp-)Cl2]2 (dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer) is an organometallic rhodium(III) complex supplied as an orange to reddish brown solid for research use in organometallic chemistry and catalysis. It serves as a catalyst precursor or reagent in synthetic and mechanistic studies and is provided with batch-level analytical data and storage recommendations.
- High purity: 98.2% as reported on the certificate of analysis.
- Contains 33.0% rhodium by mass.
- Molecular formula C20H30Cl4Rh2 and molar mass 618.08 g·mol⁻¹.
- Orange to reddish brown solid appearance.
- Suitable as a precursor or catalyst in organometallic synthesis.
- Supplied with a certificate of analysis for batch confirmation.
- Store at 4°C, protect from light, and keep under inert atmosphere; in solution: -80°C (up to 6 months) or -20°C (up to 1 month).
- Available in multiple laboratory pack sizes to support research workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.