Learn More
Medchemexpress LLC Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer | 12354-85-7 | MFCD00061552 | 98.2% | 618.08 g·mol⁻¹ | C20H30Cl4Rh2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW019734500MG
Encompass_Preferred
Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer is a dinuclear organometallic reagent, commonly represented as [(C5Me5)RhCl2]2, used as a precursor to Cp*Rh complexes and in homogeneous catalysis and synthetic organometallic chemistry. It is supplied as a solid and is used for catalyst preparation, ligand transfer, and research-scale catalytic reactions.
  • High-purity solid suitable for synthetic applications.
  • Serves as a precursor to pentamethylcyclopentadienyl rhodium complexes.
  • Facilitates formation of mononuclear rhodium catalytic species.
  • Stable under recommended storage conditions.
  • Useful for research and homogeneous catalysis workflows.
Catalog No. 50-003-01499