Learn More
Medchemexpress LLC Dichloroxylenol | 133-53-9 | MFCD00019981 | 95.4% | 191.05 g/mol | C8H8Cl2O | 100 G
SDP
Supplier:  Medchemexpress LLC HYW140799100G
Encompass_Preferred
Dichloroxylenol (2,4-dichloro-3,5-dimethylphenol; CAS 133-53-9) is a bactericide used as a preservative and deodorant for research applications. It is supplied as a research-grade chemical (molecular formula C8H8Cl2O) and is reported for use in studies such as endometritis and cervicitis in Holstein cows. Typical purity is 95.44% and it is offered in multiple package sizes for laboratory use.
  • Bactericide, preservative, and deodorant properties.
  • Reported application in reproductive tract research in cattle.
  • Research-grade purity (95.44%).
  • Available in multiple package sizes for lab convenience.
  • Molecular formula C8H8Cl2O and molecular weight 191.05 g/mol.
Catalog No. 50-002-82875