Learn More
Apexbio Technology LLC Dichlorphenamide 120-97-8 50mg
SDP
Supplier:  Apexbio Technology LLC A840850
Encompass_Preferred
Dichlorphenamide (CAS 120-97-8) is a small molecule inhibitor of carbonic anhydrase an enzyme that catalyzes the reversible hydration of carbon dioxide to bicarbonate and protons By inhibiting carbonic anhydrase dichlorphenamide modulates intracellular and extracellular pH homeostasis This compound is used in biomedical research to investigate physiological processes dependent on carbonic anhydrase activity such as acid-base balance ion transport and cellular respiration Its application facilitates the study of metabolic pathways and disorders associated with dysregulated carbonic anhydrase function
Catalog No. 50-000-94029