Learn More
Medchemexpress LLC Diclazuril | 101831-37-2 | 99.7% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0357100MG
Encompass_Preferred
Diclazuril (R-64433) is a potent and orally active anticoccidial agent, a benzeneacetonitrile derivative. It is used in the research of certain infectious and parasitic diseases.
  • Potent and orally active anticoccidial agent
  • Used in research for infectious and parasitic diseases
  • Effective against coccidiosis, acute toxoplasmosis, and equine protozoal pyoencephalitis (EPM)
Catalog No. 50-004-06114