Learn More
Medchemexpress LLC Diclofenac-13C6 | 1261393-71-8 | C813C6H11Cl2NO2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY15036S11MG
Encompass_Preferred
Diclofenac-13C6 is a stable isotope-labeled version of Diclofenac, a potent and nonselective anti-inflammatory agent and COX inhibitor. It exhibits IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells, respectively, and 5.1 and 0.84 μM for ovine COX-1 and COX-2. This compound also induces apoptosis of neural stem cells (NSCs) by activating the caspase cascade. It is intended for research use only.
  • Stable isotope-labeled Diclofenac
  • Potent and nonselective anti-inflammatory agent
  • Acts as a COX inhibitor
  • Induces apoptosis of neural stem cells (NSCs)
  • Can be used as a tracer or internal standard for quantitative analysis
  • Not a hazardous substance or mixture
  • Stable under recommended storage conditions
  • Appearance: Solid
Catalog No. 50-003-75151