Learn More
Medchemexpress LLC Diclofenac acid | 15307-86-5 | C14H11Cl2NO2 | 10 G
SDP
Supplier:  Medchemexpress LLC HY1503610G
Encompass_Preferred
Diclofenac is a potent and nonselective anti-inflammatory agent. It acts as a COX inhibitor, with IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells, and 5.1 and 0.84 μM for ovine COX-1 and COX-2, respectively. It also induces apoptosis of neural stem cells (NSCs) via the activation of the caspase cascade.
  • Targets COX
  • Induces apoptosis of neural stem cells (NSCs)
  • Supports immunology and inflammation research
  • Suitable for laboratory and R&D use
  • Available as a crystalline powder
Catalog No. 50-003-75528