Learn More
Cayman Chemical DIclofenac amIde 500mg
SDP
Supplier:  Cayman Chemical 21969500 MG
Encompass_Preferred
A prodrug form of diclofenac; decreases paw thickness in a rat model of carrageenan-induced paw edema at 100 μmol/kg; decreases acetic acid-induced writhing in mice at 100 μmol/kg; has reduced ulcerogenicity in rats compared to diclofenac; a degradation product of diclofenac resulting from photo-transformation or spontaneous cyclization in strongly acidic pH conditions; has been used as a synthetic intermediate in the synthesis of compounds with anticancer activity as well as compounds with COX-1, COX-2, and/or 5-LO inhibitory activity
Catalog No. 50-298-7568