Learn More
Medchemexpress LLC Diclofenac-d4 | 153466-65-0 | C14H7D4Cl2NO2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY15036S1MG
Encompass_Preferred
Diclofenac-d4 is the deuterium labeled form of Diclofenac, a potent and nonselective anti-inflammatory COX inhibitor. It also induces apoptosis in neural stem cells. This compound is valuable as a tracer and internal standard for quantitative analysis, leveraging the impact of deuterium incorporation on drug pharmacokinetic and metabolic profiles.
  • Potent, nonselective anti-inflammatory agent
  • Acts as a COX inhibitor
  • Induces apoptosis in neural stem cells (NSCs)
  • Suitable for use as a tracer
  • Internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Appears as a white to off-white solid
  • Soluble in ethanol (≥ 30 mg/mL) and DMSO (≥ 30 mg/mL)
  • For research use only
Catalog No. 50-003-75256