Learn More
Selleck Chemical LLC Diclofenac Diethylamine S3063-50mg
SDP
Supplier:  Selleck Chemical LLC S306350MG
Encompass_Preferred
Diclofenac diethylamine is a nonsteroidal anti-inflammatory drug taken to reduce inflammation and as an analgesic reducing pain in certain conditions Diclofenac exerts its action via inhibition of prostaglandin synthesis by inhibiting cyclooxygenase-1 (COX-1) and cyclooxygenase-2 (COX-2) with relative equipotency
Catalog No. 50-310-9218