Learn More
Medchemexpress LLC Diclofenac epolamine | 119623-66-4 | 99.9% | 411.32 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10954710MG
Encompass_Preferred
Diclofenac epolamine is a non-steroidal anti-inflammatory drug (NSAID). It is used for relieving arthritis pain, acute pain, osteoarthritis, and actinic keratosis. It has good skin absorption characteristics without local adverse reactions and allergies.
  • Acts as a non-steroidal anti-inflammatory drug (NSAID)
  • Used for relieving arthritis pain, acute pain, osteoarthritis, and actinic keratosis
  • Provides good skin absorption characteristics without local adverse reactions and allergies
  • Increases diclofenac partitioning into the skin
  • Enhances the transport of drug through skin and sustains for 12 hours after removal of microemulsion (ME)
  • Effective in treating inflammatory conditions
Catalog No. 50-003-45407