Learn More
Medchemexpress LLC Diclofenac ethyl ester | 15307-77-4 | 99.3% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12712510MM/1ML
Encompass_Preferred
Diclofenac ethyl ester is the prodrug of Diclofenac. It is encapsulated in polymer micelles in vitro and released with prolonged circulation time, leading to reduced drug concentrations in the heart and kidneys in vivo.
  • Prodrug of Diclofenac
  • Encapsulated in polymer micelles in vitro
  • Released with prolonged circulation time
  • Reduces drug concentrations in the heart and kidneys in vivo
  • For research use only
Catalog No. 50-003-42647