Learn More
Medchemexpress LLC Diclofenac ethyl ester | 15307-77-4 | MFCD17170099 | 99.3% | 324.20 g/mol | C16H15Cl2NO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1271255MG
Encompass_Preferred
Diclofenac ethyl ester is the ethyl-ester prodrug of diclofenac used for research on prodrug behavior and controlled-release formulations. Reports describe prolonged circulation and reduced cardiac and renal exposure when encapsulated in polymer micelles. Chemical identifiers include CAS 15307-77-4, formula C16H15Cl2NO2, and molecular weight 324.20 g/mol.
  • Ethyl-ester prodrug of diclofenac.
  • Used to study controlled-release and polymer micelle delivery systems.
  • Reported to prolong circulation and reduce heart and kidney exposure in vivo.
  • Well-defined chemical properties: C16H15Cl2NO2, 324.20 g/mol.
  • Supplied in small milligram quantities for laboratory research.
Catalog No. 50-002-63975