Learn More
Cayman Chemical DIclofenac ethyl ester 50mg
SDP
Supplier:  Cayman Chemical 2210550 MG
Encompass_Preferred
An esterified form of diclofenac; has been used as a precursor in the synthesis of diclofenac derivatives with anti-inflammatory and analgesic activity and decreased ulcerogenicity
Catalog No. 50-296-4180