Learn More
AdipoGen Diclofenac Impurity B
SDP
Supplier:  AdipoGen CDXD1228M050
Encompass_Preferred
Chemical. CAS 22121-58-0. Formula C13H9Cl2NO. MW 266.12. Diclofenac Impurity B is a degradation product of and a regulatory impurity in the drug Diclofenac which is a non-steroidal anti-inflammatory drug NSAID that is clinically used to treat inflammation associated with arthritis and gout as well as other pain or inflammatory disorders. This product can be used as a working standard or secondary analytical reference standard. It can also be used as an organic building block and intermediate for synthesis.
Catalog No. 50-256-1008