Learn More
Medchemexpress LLC Diclofenac methyl es 25mg | 15307-78-5 | 310.18 g/mol | 25 MG

Supplier: Medchemexpress LLC HY11468925MG
Diclofenac methyl ester (CAS 15307-78-5) is the methyl ester metabolite of diclofenac provided as a high-purity research reagent for biochemical, toxicological, and analytical studies. The material is supplied in small research packs for use as a reference standard and reagent; users should consult the datasheet and SDS for safety and handling details.
- Provided as a purified reference material suitable for laboratory research.
- Purity ≥98.04% as listed by the supplier.
- Molecular formula C15H13Cl2NO2; molecular weight 310.18 g/mol.
- 25 mg pack size suitable for method development and analytical work.
- Intended for research use only; not for human or veterinary use.
- Consult the product datasheet and SDS for storage and handling instructions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.