Learn More
Cayman Chemical DIclofenac methyl ester 100mg
SDP
Supplier:  Cayman Chemical 22218100 MG
Encompass_Preferred
A prodrug form of diclofenac; more soluble than diclofenac acid in isopropylmyristate; does not permeate human epidermal membranes in vitro
Catalog No. 50-298-6358