Learn More
Medchemexpress LLC Diclofenac potassium | 15307-81-0 | C14H10Cl2KNO2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1503810MM/1ML
Encompass_Preferred
Diclofenac potassium is a potent and nonselective anti-inflammatory agent that acts as a COX inhibitor.
  • Potent and nonselective anti-inflammatory agent.
  • Acts as a COX inhibitor.
  • Induces apoptosis of neural stem cells (NSCs) via activation of the caspase cascade.
  • Effectively blocks COX-1 mediated prostanoid production.
  • Increases expression of cleaved (activated) caspase-3.
  • Significantly increases fecal 51Cr excretion.
  • Demonstrates in vivo anti-inflammatory activity.
Catalog No. 50-003-75840