Learn More
Medchemexpress LLC Diclofenac Sodium | 15307-79-6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1503710MM/1ML
Encompass_Preferred
Diclofenac Sodium is a potent and nonselective anti-inflammatory agent that functions as a COX inhibitor. It demonstrates inhibitory activity against human COX-1 and COX-2, and against ovine COX-1 and COX-2.
  • Potent and nonselective anti-inflammatory agent
  • Functions as a COX inhibitor
  • Inhibits human COX-1 and COX-2 with IC50 values of 4 nM and 1.3 nM, respectively, in CHO cells
  • Inhibits ovine COX-1 and COX-2 with IC50 values of 5.1 μM and 0.84 μM
  • Can induce apoptosis in neural stem cells through activation of the caspase cascade
Catalog No. 50-003-75198