Learn More
Medchemexpress LLC Diclofenac Sodium | 15307-79-6 | C14H10Cl2NNaO2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15037R100MG
Encompass_Preferred
Diclofenac Sodium (Standard) is an analytical standard intended for research and analytical applications. It functions as a potent and nonselective anti-inflammatory agent and a COX inhibitor, which can also induce apoptosis of neural stem cells via caspase cascade activation.
  • Analytical standard for research and analytical applications
  • Potent and nonselective anti-inflammatory agent
  • COX inhibitor
  • Induces apoptosis of neural stem cells via caspase cascade activation
  • Used as a reference standard for assays
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
Catalog No. 50-003-75837