Learn More
Selleck Chemical LLC Diclofenac Sodium S1903-200mg
SDP
Supplier:  Selleck Chemical LLC S1903200MG
Encompass_Preferred
Diclofenac Sodium (GP 45840) is a non-selective COX inhibitor with IC50 of 0 5 g/ml and 0 5 g/ml for COX-1 and -2 in intact cells respectively used as a nonsteroidal anti-inflammatory drug (NSAID) to relieve pain and reduce swelling in flammation
Catalog No. 50-310-9775