Learn More
Medchemexpress LLC Diclofenac (Standard) | 15307-86-5 | C₁₄H₁₁Cl₂NO₂ | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15036R100MG
Encompass_Preferred
Diclofenac (Standard) is an analytical standard intended for research and analytical applications. It functions as a potent, nonselective anti-inflammatory agent and a COX inhibitor. It also induces apoptosis of neural stem cells (NSCs) through the activation of the caspase cascade.
  • Analytical standard for research and analytical applications
  • Potent and nonselective anti-inflammatory agent
  • COX inhibitor
  • Induces apoptosis of neural stem cells (NSCs)
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS
Catalog No. 50-003-75930