Learn More
Medchemexpress LLC Diclofenac (Standard) | 15307-86-5 | C14H11Cl2NO2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15036R50MG
Encompass_Preferred
Diclofenac (Standard) is the analytical standard of Diclofenac and is intended for research and analytical applications. It is a potent and nonselective anti-inflammatory agent that acts as a COX inhibitor. Diclofenac induces apoptosis of neural stem cells (NSCs) via the activation of the caspase cascade.
  • Analytical standard
  • Reference standard for assays
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-003-75255