Learn More
Research Products International Corp Dicloxacillin Sodium Salt 1 G
SDP
Supplier:  Research Products International Corp D821501.0
Encompass_Preferred
Dicloxacillin sodium salt is a narrow-spectrum antibiotic in the penicillin class, specifically belonging to the beta-lactamase-resistant penicillins.
As a beta-lactam antibiotic, dicloxacillin works by inhibiting the synthesis of bacterial cell walls, leading to the destruction of the bacterial cell. 
Catalog No. 50-223-0525