Learn More
Medchemexpress LLC Dicloxacillin sodium | 343-55-5 | 98.0% | 492.31 | C19H16Cl2N3NaO5S | 10 MG

Supplier: Medchemexpress LLC HYB145910MG
Dicloxacillin sodium is a β-lactam antibiotic of the penicillin class supplied for research and analytical applications. It is active against gram-positive bacteria, including β-lactamase-producing Staphylococcus aureus, and is offered in small milligram quantities for laboratory use.
- β-lactam antibiotic effective against gram-positive bacteria
- 98.0% reported purity
- available in multiple sizes, including 10 MG
- molecular formula C19H16Cl2N3NaO5S; molecular weight 492.31
- CAS number 343-55-5
- intended for research and analytical use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.