Learn More
Medchemexpress LLC Dicloxacillin sodium | 343-55-5 | 98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB145950MG
Encompass_Preferred
Dicloxacillin sodium is a β-lactam antibiotic belonging to the penicillin family. It is effective against Gram-positive bacteria, including β-lactamase-producing organisms such as *Staphylococcus aureus*.
  • Active against Gram-positive bacteria.
  • Effective against β-lactamase-producing organisms.
  • A β-lactam antibiotic.
  • Belongs to the penicillin family.
Catalog No. 50-004-13354