Learn More
Alkali Scientific Dicloxacillin Sodium Salt, 1 G
SDP
Supplier:  Alkali Scientific DIC002CP
Encompass_Preferred
Dicloxacillin Sodium Salt, 1g, is an antibiotic used in laboratory research to study its effects on bacterial infections. It is particularly effective against penicillin-resistant Gram-positive bacteria, including strains of Staphylococcus aureus. In microbiological experiments, Dicloxacillin is used to test bacterial resistance and evaluate new antimicrobial agents. The 1g volume allows for more extensive testing, making it suitable for larger-scale experiments or studies where higher dosages are required. Dicloxacillin is frequently used in combination therapy research to enhance the effectiveness of other antibiotics in combating bacterial resistance.
Catalog No. 50-006-60209